Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beta-Galactosidase Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89138051 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-051 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-051 Supplier Abnova Corporation Supplier No. PAB15376

Rabbit polyclonal antibody raised against native Beta-Galactosidase.

Specifications

Antigen Beta-Galactosidase
Applications ELISA, Immunodiffusion, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation In 20mM sodium phosphate buffer, 140mM NaCl, pH 7.3
Host Species Rabbit
Immunogen Native purified Escherichia coli Beta-Galactosidase.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity By immunoelectrophoresis, This antibody gives a single precipitin arc against anti-rabbit serum and purified and partially purified b-galactosidase (E. coli). Cross-reactivity from other tissues and species may occur. This product is suitable for use as a precipitating antibody. This product is specially prepared for the detection and purification of recombinant proteins. It shows reactivity to beta-galactosidase and is tested by Western or dot blot to ensure monospecificity using lysates of D-lac and strain 3.3 of E. coli. No reaction to other E. Coli proteins detected.
Target Species E. coli
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.