Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIRC4, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114103 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-103 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-103 Supplier Abnova Corporation Supplier No. PAB0282
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant BIRC4.

The protein encoded by this gene is a member of a family of proteins which inhibit apoptosis through binding to tumor necrosis factor receptor-associated factors TRAF1 and TRAF2. This protein inhibits apoptosis induced by menadione, a potent inducer of free radicals, and ICE. It also inhibits at least two members of the caspase family of cell-death proteases, caspase-3 and caspase-7. [provided by RefSeq

Specifications

Antigen BIRC4
Applications Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant BIRC4.
Dilution The optimal working dilution should be determined by the end user.
Formulation In serum (0.05% sodium azide)
Gene XIAP
Gene Alias API3/BIRC4/ILP1/MIHA/XLP2
Gene Symbols XIAP
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 163-230 of human BIRC4.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 331
Target Species Gerbil, Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.