Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIRC5 Rabbit Biotin Polyclonal Antibody Abnova™ (phospho S20), Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89117484 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-484 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-484 Supplier Abnova Corporation Supplier No. PAB15182
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of BIRC5.

This gene is a member of the inhibitor of apoptosis (IAP) gene family, which encode negative regulatory proteins that prevent apoptotic cell death. IAP family members usually contain multiple baculovirus IAP repeat (BIR) domains, but this gene encodes proteins with only a single BIR domain. The encoded proteins also lack a C-terminus RING finger domain. Gene expression is high during fetal development and in most tumors yet low in adult tissues. Antisense transcripts are involved in the regulation of this gene's expression. At least four transcript variants encoding distinct isoforms have been found for this gene, but the full-length natures of only three of them have been determined. [provided by RefSeq]

Specifications

Antigen BIRC5
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of BIRC5.
Dilution Western Blot (3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene BIRC5
Gene Accession No. O15392
Gene Alias API4/EPR-1
Gene Symbols BIRC5
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding S20 of human BIRC5.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 332
Test Specificity This antibody is useful for Western blot, where a band is seen ∼16.5 KDa.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.