Learn More
BIRC5 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BIRC5.
Supplier: Abnova Corporation PAB9042
Description
This gene is a member of the inhibitor of apoptosis (IAP) gene family, which encode negative regulatory proteins that prevent apoptotic cell death. IAP family members usually contain multiple baculovirus IAP repeat (BIR) domains, but this gene encodes proteins with only a single BIR domain. The encoded proteins also lack a C-terminus RING finger domain. Gene expression is high during fetal development and in most tumors yet low in adult tissues. Antisense transcripts are involved in the regulation of this gene's expression. At least four transcript variants encoding distinct isoforms have been found for this gene, but the full-length natures of only three of them have been determined. [provided by RefSeq]
Specifications
BIRC5 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of BIRC5. | |
In PBS, pH 7.2 (30% glycerol, 0.5mg/mL BSA, 0.01% thimerosal) | |
API4/EPR-1 | |
Rabbit | |
100 μg | |
Primary | |
Human | |
Liquid |
Immunofluorescence, Western Blot | |
Unconjugated | |
Western Blot (0.5-4 ug/mL) The optimal working dilution should be determined by the end user. | |
BIRC5 | |
BIRC5 | |
A synthetic peptide corresponding to C-terminus of human BIRC5. | |
RUO | |
332 | |
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only