Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIRC7 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89117232 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-232 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-232 Supplier Abnova Corporation Supplier No. PAB14670
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of BIRC7.

The protein encoded by this gene is a member of the family of inhibitor of apoptosis proteins (IAP) and contains a single copy of a baculovirus IAP repeat (BIR) as well as a RING-type zinc finger domain. The BIR domain is essential for inhibitory activity and interacts with caspases, while the RING finger domain sometimes enhances antiapoptotic activity but does not inhibit apoptosis alone. Two transcript variants encoding different isoforms have been found for this gene. The two isoforms have different antiapoptotic properties, with isoform alpha protecting cells from apoptosis induced by staurosporine and isoform b protecting cells from apoptosis induced by etoposide. [provided by RefSeq

Sequence: PELPTPRREVQSESAQEPG

Specifications

Antigen BIRC7
Applications Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of BIRC7.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000-1:5000) Immunoprecipitation (1:50-1:200) Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user.
Formulation In serum (0.05% sodium azide)
Gene BIRC7
Gene Accession No. NP_647478;NP_071444.1
Gene Alias KIAP/LIVIN/ML-IAP/MLIAP/RNF50
Gene Symbols BIRC7
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 199-217 of human BIRC7.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79444
Test Specificity Livin is a short-lived protein that is thought to be degraded by the ubiquitin-proteasome pathway. Thus users may want to do a time course when examining the effects of treatments on Livin expression. Users are encouraged to refer to the literature for additional information; for example, Ma et al. 2006. Livin has been shown to undergo proteolytic cleavage during apoptosis. Thus the molecular weight of Livin observed on Western blots may vary depending on the presence of absence of cleavage fragments.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.