Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLCAP, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89119031 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-119-031 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-119-031 Supplier Abnova Corporation Supplier No. PAB17374
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of BLCAP.

The protein encoded by this gene was identified using a differential display procedure with tumor biopsies obtained from a noninvasive and an invasive bladder transitional cell carcinoma. Although database searches revealed no homology to any human gene at the time of identification, mouse, rat and zebrafish orthologs have since been identified. The function of this differentially expressed protein is not yet known but it appears to be down-regulated during bladder cancer progression. [provided by RefSeq

Specifications

Antigen BLCAP
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of BLCAP.
Dilution Immunofluorescence (1:500-1:1000) ELISA (1:40000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol)
Gene BLCAP
Gene Accession No. P62952
Gene Alias BC10
Gene Symbols BLCAP
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal of human BLCAP.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10904
Test Specificity This antibody detects endogenous levels of total BLCAP protein.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.