Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLOC1S1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89122890 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-122-890 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-122-890 Supplier Abnova Corporation Supplier No. PAB21399
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant BLOC1S1.

BLOC1S1 is a component of the ubiquitously expressed BLOC1 multisubunit protein complex. BLOC1 is required for normal biogenesis of specialized organelles of the endosomal-lysosomal system, such as melanosomes and platelet dense granules (Starcevic and Dell'Angelica, 2004 [PubMed 15102850]).[supplied by OMIM

Sequence: LLKEHQAKQNERKELQEKRRREAITAATCLTEALVDHLNVGVAQAYMNQRKLDHEVKTLQVQAAQFAKQTGQWIGMVENFNQALKEIGDVENWARSIELDMRTIATALEYVYKGQ

Specifications

Antigen BLOC1S1
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant BLOC1S1.
Dilution Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene BLOC1S1
Gene Accession No. P78537
Gene Alias BLOS1/FLJ39337/FLJ97089/GCN5L1/MGC87455/MICoA/RT14
Gene Symbols BLOC1S1
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human BLOC1S1.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2647
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.