Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BNIP3 Rabbit anti-Chicken, Clawed frog, Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114086 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-086 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-086 Supplier Abnova Corporation Supplier No. PAB0264
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of BNIP3.

This gene is a member of the BCL2/adenovirus E1B 19kDa-interacting protein (BNIP) family. It interacts with the E1B 19kDa protein which is responsible for the protection of virally-induced cell death, as well as E1B 19kDa-like sequences of BCL2, also an apoptotic protector. This gene contains a BH3 domain and a transmembrane domain, which have been associated with pro-apoptotic function. The dimeric mitochondrial protein encoded by this gene is known to induce apoptosis, even in the presence of BCL2. [provided by RefSeq

Sequence: SRPENIPPKEFLFKHPKR

Specifications

Antigen BNIP3
Applications Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of BNIP3.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000-1:5000) Immunohistochemistry (Frozen sections) Immunoprecipitation (1:50-1:200) Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the
Formulation In serum (0.05% sodium azide)
Gene BNIP3
Gene Alias NIP3
Gene Symbols BNIP3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 122-139 of human BNIP3.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 664
Target Species Chicken, Frog, Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.