Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BNIP3L, BNIP3, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89120186 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-186 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-186 Supplier Abnova Corporation Supplier No. PAB1863
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of BNIP3 (BH3 Domain Specific).

This gene is a member of the BCL2/adenovirus E1B 19kDa-interacting protein (BNIP) family. It interacts with the E1B 19kDa protein which is responsible for the protection of virally-induced cell death, as well as E1B 19kDa-like sequences of BCL2, also an apoptotic protector. This gene contains a BH3 domain and a transmembrane domain, which have been associated with pro-apoptotic function. The dimeric mitochondrial protein encoded by this gene is known to induce apoptosis, even in the presence of BCL2. [provided by RefSeq

Specifications

Antigen BNIP3L, BNIP3
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of BNIP3 (BH3 Domain Specific).
Dilution Western Blot (1:1000) Immunohistochemistry (1:50-100) Immunofluorescence (1:50-100) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene BNIP3
Gene Accession No. NP_004322 (Gene ID : 665);Q12983 (Gene ID : 664)
Gene Alias NIP3
Gene Symbols BNIP3
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human BNIP3L, BNIP3.
Purification Method Protein G purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 664
Test Specificity BH3 Domain Specific
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.