Learn More
Rabbit anti-Bovine Albumin, Polyclonal Antibody, MP Biomedicals
Shop All MP Biomedicals, Inc ProductsDescription
Host Species: Rabbit
Species Reactivity: Bv
Immunogen: Fraction V bovine serum albumin
Immunoassays, Western Blotting
Specifications
Specifications
| Antigen | Bovine Albumin |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | Fraction V bovine serum albumin |
| Purification Method | Affinity Purified |
| Quantity | 2 mg |
| Primary or Secondary | Primary |
| Target Species | Bovine |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.