Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Bovine IgG F(ab')2 Secondary Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NBP173273 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP173273 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP173273 Supplier Novus Biologicals Supplier No. NBP173273
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IgG F(ab')2 Polyclonal antibody specifically detects IgG F(ab')2 in Bovine samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA.

Specifications

Antigen IgG F(ab')2
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration LYOPH
Conjugate Fluorescein
Dilution Flow Cytometry 1:1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000
Gene Alias immunoglobulin heavy constant gamma 1 (G1m marker)
Host Species Rabbit
Immunogen Bovine IgG F(ab')2 fragment
Purification Method Affinity Purified
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity No reaction was observed against Bovine IgG F(c).
Reconstitution Reconstitute with 1.0 ml deionized water (or equivalent).
Target Species Bovine
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.