Learn More
BRAF Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BRAF.
Supplier: Abnova Corporation PAB3946
Description
This gene encodes a protein belonging to the raf/mil family of serine/threonine protein kinases. This protein plays a role in regulating the MAP kinase/ERKs signaling pathway, which affects cell division, differentiation, and secretion. Mutations in this gene are associated with cardiofaciocutaneous syndrome, a disease characterized by heart defects, mental retardation and a distinctive facial appearance. Mutations in this gene have also been associated with various cancers, including non-Hodgkin lymphoma, colorectal cancer, malignant melanoma, thyroid carcinoma, nonsmall cell lung carcinoma, and adenocarcinoma of lung. A pseudogene, which is located on chromosome X, has been identified for this gene. [provided by RefSeq
Specifications
BRAF | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of BRAF. | |
In PBS (0.09% sodium azide) | |
P15056 | |
BRAF | |
A synthetic peptide (conjugated with KLH) corresponding to residues surrounding T400 of human BRAF. | |
400 μL | |
Primary | |
Human | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:10-50) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
BRAF | |
B-RAF1/BRAF1/FLJ95109/MGC126806/MGC138284/RAFB1 | |
Rabbit | |
Protein A purification | |
RUO | |
673 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only