Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUB1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89116519 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-519 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-519 Supplier Abnova Corporation Supplier No. PAB13270
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of BUB1.

This gene encodes a kinase involved in spindle checkpoint function. The kinase functions in part by phosphorylating a member of the miotic checkpoint complex and activating the spindle checkpoint. Mutations in this gene have been associated with aneuploidy and several forms of cancer. [provided by RefSeq

Specifications

Antigen BUB1
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of BUB1.
Dilution Western Blot (2-4 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene BUB1
Gene Accession No. O43683
Gene Alias BUB1A/BUB1L/hBUB1
Gene Symbols BUB1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to N-terminus 19 amino acids of human BUB1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 699
Target Species Human, Mouse
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.