Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C10orf26, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89124840 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-124-840 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-124-840 Supplier Abnova Corporation Supplier No. PAB23141
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant C10orf26.

Sequence: PLSEPSRSSTRPPSIADPDPSDLPVDRAATKAPGMEPSGSVAGLGELDPGAFLDKDAECREELLKDDSSEHGAPDSKEKTPGRHRRFTGD

Specifications

Antigen C10orf26
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant C10orf26.
Dilution Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene C10orf26
Gene Accession No. Q9NX94
Gene Alias FLJ20154/FLJ20367/MGC125648/OPAL1
Gene Symbols C10orf26
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human C10orf26.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54838
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.