Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C10orf68, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89125110 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-125-110 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-125-110 Supplier Abnova Corporation Supplier No. PAB23411

Rabbit polyclonal antibody raised against recombinant C10orf68.

Sequence: ENPMLKHQDSVSKIQVQLEIQETSEGEGRSIPDKNSMFVHQDSVSKLQMQEKKKITPGRERRNTRIVVPNENVISVHQDSKSKLQMQEKKQINSGV

Specifications

Antigen C10orf68
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description chromosome 10 open reading frame 68
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene C10orf68
Gene Accession No. Q9H943
Gene Alias FLJ13031, MGC149767, dJ1104A8.1
Gene Symbols C10orf68
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human C10orf68.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79741
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.