Learn More
C13orf34 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of C13orf34.
Supplier: Abnova Corporation PAB16722
Description
BORA is an activator of the protein kinase Aurora A (AURKA; MIM 603072), which is required for centrosome maturation, spindle assembly, and asymmetric protein localization during mitosis (Hutterer et al., 2006 [PubMed 16890155]).[supplied by OMIM
Specifications
C13orf34 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of C13orf34. | |
In PBS (0.02% sodium azide) | |
Q6PGQ7 | |
C13orf34 | |
A synthetic peptide corresponding to C-terminus 15 amino acids from of human C13orf34. | |
RUO | |
79866 | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
C13orf34 | |
BORA/FLJ22624 | |
Rabbit | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only