Learn More
C17orf70 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant C17orf70.
Supplier: Abnova Corporation PAB21804
Description
FAAP100 is a component of the Fanconi anemia (FA; MIM 277650) core complex and is required for core complex stability and FANCD2 (see MIM 227646) monoubiquitination (Ling et al., 2007 [PubMed 17396147]).[supplied by OMIM
Sequence: VLCSVSPSGSRVPHDLLGGSGGFTLEDALFGLLFGADATLLQSPVVLCGLPDGQLCCVILKALVTSRSAPGDPNALVKILHHLEEPVIFIGALKTEPQAAEAAENFLPDEDVHCDCSpecifications
C17orf70 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant C17orf70. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
Q0VG06 | |
C17orf70 | |
Recombinant protein corresponding to amino acids of human C17orf70. | |
100 μL | |
Primary | |
Human | |
Liquid |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
C17orf70 | |
FAAP100/FLJ22175/FLJ30151 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
80233 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only