Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1orf100 Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89124232 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-124-232 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-124-232 Supplier Abnova Corporation Supplier No. PAB22523

Rabbit polyclonal antibody raised against recombinant C1orf100.

Sequence: MTAIRLREFIERRPVIPPSIFIAHQGRDVQGYYPGQLARLHFDHSAKRAPRPLIDLTIPPKTKYHYQPQLDQ

Specifications

Antigen C1orf100
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Description chromosome 1 open reading frame 100
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene C1orf100
Gene Accession No. Q5SVJ3
Gene Symbols C1orf100
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human C1orf100.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200159
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.