Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C5orf37, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89124824 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-124-824 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-124-824 Supplier Abnova Corporation Supplier No. PAB23124
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant C5orf37.

Sequence: DVVERACQARAEEVCIQISNDYEAKVAMLSGALENAKAEIQRMQHEKEHFEDSMKKAFMRGVCALNLEAMTIFQNRNDAGIDSTNNKKEEYGPGVQ

Specifications

Antigen C5orf37
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant C5orf37.
Dilution Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene C5orf37
Gene Accession No. Q8NA72
Gene Alias FLJ35779/MGC120442/MGC120443/MGC120444
Gene Symbols C5orf37
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human C5orf37.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134359
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.