Learn More
CA9 Rabbit anti-Dog, Human, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CA9.
Supplier: Abnova Corporation PAB12466
Description
Carbonic anhydrases (CAs) are a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. They participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. They show extensive diversity in tissue distribution and in their subcellular localization. CA IX is a transmembrane protein and the only tumor-associated carbonic anhydrase isoenzyme known. It is expressed in all clear-cell renal cell carcinoma, but is not detected in normal kidney or most other normal tissues. It may be involved in cell proliferation and transformation. This gene was mapped to 17q21.2 by fluorescence in situ hybridization, however, radiation hybrid mapping localized it to 9p13-p12. [provided by RefSeq
Specifications
CA9 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of CA9. | |
In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.09% sodium azide) | |
Q16790 | |
CA9 | |
A synthetic peptide corresponding to C-terminus of human CA9. | |
RUO | |
768 | |
Canine, Human, Rat | |
Liquid |
Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot | |
Unconjugated | |
Immunofluorescence (1:1000) Immunohistochemistry (Frozen sections) (1:1000) Western Blot (1:2000) The optimal working dilution should be determined by the end user. | |
CA9 | |
CAIX/MN | |
Rabbit | |
100 μL | |
Primary | |
This antibody is specific to human CA IX. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only