Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CABP5, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89125755 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-125-755 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-125-755 Supplier Abnova Corporation Supplier No. PAB24053
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant CABP5.

The product of this gene belongs to a subfamily of calcium binding proteins, which share similarity to calmodulin. Calcium binding proteins are an important component of calcium mediated cellular signal transduction. Expression of this gene is retina-specific. The mouse homolog of this protein has been shown to express in the inner nuclear layer of the retina, suggested its role in neuronal functioning. The specific function of this gene is unknown. Study of the transcripts and genomic structure revealed that the 5' end of this gene is complementary and reverse to that of CABP3 gene, and the sequence beyond the overlapping region is nearly identical to that of CABP3. Thus, these two genes encode the protein products with distinct N-terminal halves but identical C-terminal halves. [provided by RefSeq]

Sequence: ITLVELQQAMQRLLGERLTPREISEVVREADVNGDG

Specifications

Antigen CABP5
Applications Immunohistochemistry, Western Blot, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant CABP5.
Dilution Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene CABP5
Gene Accession No. Q9NP86
Gene Symbols CABP5
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human CABP5.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56344
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.