Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CACNA1B, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89126028 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-126-028 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-126-028 Supplier Abnova Corporation Supplier No. PAB24318
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant CACNA1B.

Voltage-dependent Ca(2+) channels are multisubunit complexes found in the membrane of many excitable cells that regulate calcium entry (see MIM 601011). N-type calcium channels, which control neurotransmitter release from neurons, are dihydropyridine-insensitive and omega-conotoxin-sensitive. The alpha-1 subunit forms the pore through which calcium enters the cell, and is encoded by a family of at least 5 genes.[supplied by OMIM

Sequence: ALMIFDFYKQNKTTRDQMQQAPGGLSQMGPVSLFHPLKATLEQTQPAVLRGARVFLRQKSSTSLSNGGAIQNQESGIKESVSWGTQRTQDAPHE

Specifications

Antigen CACNA1B
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant CACNA1B.
Dilution Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene CACNA1B
Gene Accession No. Q00975
Gene Alias BIII/CACNL1A5/CACNN/Cav2.2
Gene Symbols CACNA1B
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human CACNA1B.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 774
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.