Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cacna1c, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89134422 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-422 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-422 Supplier Abnova Corporation Supplier No. PAB9795
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Cacna1c.

Specifications

Antigen Cacna1c
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Cacna1c.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Gene Cacna1c
Gene Alias RATIVS302
Gene Symbols Cacna1c
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to rat Cacna1c.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24239
Target Species Human, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.