Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cacnb3 Rabbit anti-Rat, Polyclonal Antibody, Abnova™

Catalog No. 89134430 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-430 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-430 Supplier Abnova Corporation Supplier No. PAB9803

Rabbit polyclonal antibody raised against synthetic peptide of Cacnb3.

Specifications

Antigen Cacnb3
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description calcium channel, voltage-dependent, beta 3 subunit
Formulation In PBS, pH 7.4 (0.08% sodium azide)
Gene Cacnb3
Gene Alias CACH3B
Gene Symbols Cacnb3
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 1-15 of rat Cacnb3.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25297
Target Species Rat
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.