Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cacng2 Rabbit anti-Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Cacng2.
Supplier: Abnova Corporation PAB9618
Specifications
Cacng2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Cacng2. | |
In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 10% BSA) | |
O88602 | |
Cacng2 | |
A synthetic peptide (conjugated with KLH) corresponding to C-terminus rat Cacng2. | |
100 μL | |
Primary | |
Mouse, Rat | |
Liquid |
Western Blot | |
Unconjugated | |
Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
Cacng2 | |
Ipr328 | |
Rabbit | |
Affinity purification | |
RUO | |
84347 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction