Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cacng3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Cacng3.
Supplier: Abnova Corporation PAB9885
Specifications
Cacng3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Cacng3. | |
In PBS (0.08% sodium azide) | |
Cacng3 | |
A synthetic peptide (conjugated with KLH) corresponding to rat Cacng3. | |
RUO | |
140724 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
Western Blot (5-10 ug/mL) The optimal working dilution should be determined by the end user. | |
Cacng3 | |
Rabbit | |
100 μg | |
Primary | |
Human, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction