Learn More
CACNG8, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant CACNG8.
Supplier: Abnova Corporation PAB23789
Description
Voltage-dependent calcium channels couple membrane depolarization in a number of cellular processes. These activities are regulated by distinct channels composed of the pore-forming alpha-1 subunit and the modulatory beta, alpha-2/delta and gamma subunits. The protein encoded by this gene represents one of the gamma subunits. It is an integral membrane protein that is thought to stabilize the calcium channel in an inactive (closed) state. This gene is located in a cluster with two similar gamma subunit-encoding genes on chromosome 19. The mRNA for this gene is believed to initiate translation from a non-AUG (CUG) start codon. [provided by RefSeq
Sequence: DYWLYTRALICNTTNLTAGGDDGTPHRGGGGASEKKDPGGLTHSGLSpecifications
CACNG8 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant CACNG8. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
Q8WXS5 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
59283 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
CACNG8 | |
CACNG8 | |
Recombinant protein corresponding to amino acids of human CACNG8. | |
100 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only