Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CALHM1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89120719 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-719 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-719 Supplier Abnova Corporation Supplier No. PAB19190
Add to Cart
Add to Cart

Rabbit polyclonal antibody rasied against synthetic peptide of CALHM1.

CALHM1 is a cerebral Ca(2+) channel that controls processing of amyloid-beta (A-beta) precursor protein (APP; MIM 104760) (Dreses-Werringloer et al., 2008 [PubMed 18585350]).[supplied by OMIM

Specifications

Antigen CALHM1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody rasied against synthetic peptide of CALHM1.
Dilution Western blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene CALHM1
Gene Accession No. NP_001001412
Gene Alias FAM26C/MGC39514/MGC39617
Gene Symbols CALHM1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 17 amino acids near N-terminus of human CALHM1.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 255022
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.