Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-CAMK1, Polyclonal, Abnova™

Catalog No. 89117152 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-152 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-152 Supplier Abnova Corporation Supplier No. PAB14571

Specifications

Antigen CAMK1
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation In serum (50% glycerol, 0.05% sodium azide)
Gene Accession No. NP_003647
Gene Symbols CAMK1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 319-334 of human CAMK1.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 8536
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.