Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CANX Rabbit anti-Birds, Bovine, Chicken, Dog, Frog, Fruit fly, Guinea pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep, Polyclonal Antibody, Abnova™

Catalog No. 89117606 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-606 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-606 Supplier Abnova Corporation Supplier No. PAB15414
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CANX.

Specifications

Antigen CANX
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CANX.
Dilution Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Formulation In antiserum
Gene CANX
Gene Alias PP90
Gene Symbols CANX
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to C-terminus of dog CANX.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 403908
Test Specificity Detects ∼90KDa. Identical to human, mouse and rat calnexin sequences over these residues.
Target Species Avian, Bovine, Canine, Chicken, Drosophila, Frog, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.