Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAPRIN1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CAPRIN1.
Supplier: Abnova Corporation PAB4337
Specifications
| CAPRIN1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CAPRIN1. | |
| In PBS (0.09% sodium azide) | |
| NP_976240;Q14444 | |
| CAPRIN1 | |
| A synthetic peptide (conjugated with KLH) corresponding to amino acids 625-654 at C-terminus of human CAPRIN1. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| CAPRIN1 | |
| GPIAP1/GPIP137/M11S1/caprin-1/p137GPI | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 4076 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction