Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CARD8 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89114102 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-102 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-102 Supplier Abnova Corporation Supplier No. PAB0281
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against full-length recombinant CARD8.

The protein encoded by this gene belongs to the caspase recruitment domain (CARD)-containing family of proteins, which are involved in pathways leading to activation of caspases or nuclear factor kappa-B (NFKB). This protein may be a component of the inflammasome, a protein complex that plays a role in the activation of proinflammatory caspases. It is thought that this protein acts as an adaptor molecule that negatively regulates NFKB activation, CASP1-dependent IL1B secretion, and apoptosis. Polymorphisms in this gene may be associated with a susceptibility to rheumatoid arthritis. Alternatively spliced transcript variants have been described for this gene, but their biological validity has not been determined. [provided by RefSeq

Specifications

Antigen CARD8
Applications Immunohistochemistry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against full length recombinant CARD8.
Dilution Immunohistochemistry Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000-1:5000) Immunoprecipitation (1:50-1:200) Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user.
Formulation In serum (0.05% sodium azide)
Gene CARD8
Gene Alias CARDINAL/DACAR/DKFZp779L0366/Dakar/FLJ18119/FLJ18121/KIAA0955/MGC57162/NDPP/NDPP1/TUCAN
Gene Symbols CARD8
Host Species Rabbit
Immunogen Recombinant protein corresponding to full length human CARD8.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22900
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.