Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CASP1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89120405 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-405 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-405 Supplier Abnova Corporation Supplier No. PAB18845
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CASP1.

This gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce 2 subunits, large and small, that dimerize to form the active enzyme. This gene was identified by its ability to proteolytically cleave and activate the inactive precursor of interleukin-1, a cytokine involved in the processes such as inflammation, septic shock, and wound healing. This gene has been shown to induce cell apoptosis and may function in various developmental stages. Studies of a similar gene in mouse suggest a role in the pathogenesis of Huntington disease. Alternative splicing of this gene results in five transcript variants encoding distinct isoforms. [provided by RefSeq

Specifications

Antigen CASP1
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CASP1.
Dilution ELISA (1:40000-1:80000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:100) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.02% sodium azide, 2% BSA
Gene CASP1
Gene Accession No. NM_033292
Gene Alias ICE/IL1BC/P45
Gene Symbols CASP1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids at N-terminus of human CASP1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 834
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.