Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CASP9 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89126518 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-126-518 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-126-518 Supplier Abnova Corporation Supplier No. PAB24846
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CASP9.

This gene encodes a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. This protein is processed by caspase APAF1; this step is thought to be one of the earliest in the caspase activation cascade. Alternative splicing results in two transcript variants which encode different isoforms. [provided by RefSeq

Specifications

Antigen CASP9
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CASP9.
Dilution Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In 1X PBS, pH 7.2 (0.09% sodium azide)
Gene CASP9
Gene Alias APAF-3/APAF3/CASPASE-9c/ICE-LAP6/MCH6
Gene Symbols CASP9
Host Species Rabbit
Immunogen A synthetic peptide corresponding to CASP9.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 842
Test Specificity CASP9 polyclonal antibody detects endogenous levels of CASP9 protein.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.