Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBLC Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89118776 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-776 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-776 Supplier Abnova Corporation Supplier No. PAB1698
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CBLC.

CBL proteins, such as CBLC, are phosphorylated upon activation of a variety of receptors that signal via protein tyrosine kinases. Through interactions with proteins containing SRC (MIM 190090) homology-2 (SH2) and SH3 domains, CBL proteins modulate downstream cell signaling (Keane et al., 1999 [PubMed 10362357]).[supplied by OMIM

Specifications

Antigen CBLC
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CBLC.
Dilution Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene CBLC
Gene Accession No. NP_036248;Q9ULV8
Gene Alias CBL-3/CBL-SL/RNF57
Gene Symbols CBLC
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human CBLC.
Purification Method Protein G purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23624
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.