Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBX4 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89120414 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-414 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-414 Supplier Abnova Corporation Supplier No. PAB18854
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CBX4.

Specifications

Antigen CBX4
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CBX4.
Dilution ELISA (1:40000) Western Blot (1:200-1:1000) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.02% sodium azide
Gene CBX4
Gene Accession No. O00257
Gene Alias NBP16/PC2/hPC2
Gene Symbols CBX4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 15 amino acids of human CBX4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8535
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.