Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBX5 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89117654 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-654 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-654 Supplier Abnova Corporation Supplier No. PAB15580
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CBX5.

This gene encodes a highly conserved nonhistone protein, which is a member of the heterochromatin protein family. The protein is enriched in the heterochromatin and associated with centromeres. The protein has a single N-terminal chromodomain which can bind to histone proteins via methylated lysine residues, and a C-terminal chromo shadow-domain (CSD) which is responsible for the homodimerization and interaction with a number of chromatin-associated nonhistone proteins. The encoded product is involved in the formation of functional kinetochore through interaction with essential kinetochore proteins. The gene has a pseudogene located on chromosome 3. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq

Sequence: EDAENKEKETAKS

Specifications

Antigen CBX5
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CBX5.
Dilution Western Blot (1:1000-1:3000) Immunofluorescence (1:200-1:1000) The optimal working dilution should be determined by the end user.
Formulation In 120mM sodium phosphate buffer, pH 7.4 (50% glycerol)
Gene CBX5
Gene Alias HP1/HP1A
Gene Symbols CBX5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 179-191 of human CBX5.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23468
Target Species Human
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.