Learn More
CCDC102A Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsSpecifications
Specifications
| Antigen | CCDC102A |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against synthetic peptide of CCDC102A. |
| Dilution | Western Blot (1:500-1:1000) ELISA (1:5000) The optimal working dilution should be determined by the end user. |
| Formulation | In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) |
| Gene | CCDC102A |
| Gene Accession No. | Q96A19 |
| Gene Alias | MGC10992/MGC13119 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.