Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC88A (phospho S1417), Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89130682 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-682 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-682 Supplier Abnova Corporation Supplier No. PAB29246
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of human CCDC88A.

Specifications

Antigen CCDC88A
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of human CCDC88A.
Dilution Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (without mg2+ and Ca2+), (50% glycerol, 0.02% sodium azide)
Gene CCDC88A
Gene Accession No. Q3V6T2
Gene Alias APE/DKFZp686D0630/GIRDIN/GIV/GRDN/HkRP1/KIAA1212
Gene Symbols CCDC88A
Host Species Rabbit
Immunogen Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S1417 of human CCDC88A.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55704
Test Specificity This antibody detects endogenous levels of CCDC88A only when phosphorylated at serine 1417.
Target Species Human, Mouse
Content And Storage Store at -20°C.
Avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.