Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL2 Rabbit anti-Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89118484 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-484 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-484 Supplier Abnova Corporation Supplier No. PAB16617
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant CCL2.

This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils. It has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis and atherosclerosis. It binds to chemokine receptors CCR2 and CCR4. [provided by RefSeq

Specifications

Antigen CCL2
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against full length recombinant mouse Ccl2.
Dilution ELISA (0.5-2 ug/mL), detection limit 2-4 ng/mL using recombinant mouse CCL2 Immunohistochemistry (2-10 ug/mL) Western Blot (0.1-0.2 ug/mL), detection limit 1.5-3 ng/lane using recombinant mouse CCL2 The optimal working dilution should be determin
Formulation Lyophilized from PBS
Gene CCL2
Gene Accession No. P13500
Gene Alias AI323594/HC11/JE/MCAF/MCP-1/MCP1/SMC-CF/Scya2/Sigje
Gene Symbols Ccl2
Host Species Rabbit
Immunogen Recombinant protein corresponding to full length mouse Ccl2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20296
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.