Learn More
CCL3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CCL3.
Supplier: Abnova Corporation PAB25175
Description
Macrophage inflammatory protein-1 is a so-called monokine that is involved in the acute inflammatory state in the recruitment and activation of polymorphonuclear leukocytes (Wolpe et al., 1988 [PubMed 3279154]). Sherry et al. (1988) [PubMed 3058856] demonstrated 2 protein components of MIP1, called by them alpha and beta.[supplied by OMIM
Specifications
CCL3 | |
Polyclonal | |
Unconjugated | |
Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
CCL3 | |
CCL3 | |
A synthetic peptide corresponding to CCL3. | |
100 μL | |
Primary | |
CCL3 polyclonal antibody detects endogenous levels of CCL3 protein. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed) | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of CCL3. | |
In PBS, pH 7.2 (0.05% sodium azide) | |
G0S19-1/LD78ALPHA/MIP-1-alpha/MIP1A/SCYA3 | |
Rabbit | |
Affinity purification | |
RUO | |
6348 | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only