Learn More
CCR4 Rabbit anti-Human, Monkey, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CCR4.
Supplier: Abnova Corporation PAB16380
Description
The protein encoded by this gene belongs to the G-protein-coupled receptor family . It is a receptor for the CC chemokine - MIP-1, RANTES, TARC and MCP-1. Chemokines are a group of small polypeptide, structurally related molecules that regulate cell trafficking of various types of leukocytes. The chemokines also play fundamental roles in the development, homeostasis, and function of the immune system, and they have effects on cells of the central nervous system as well as on endothelial cells involved in angiogenesis or angiostasis. [provided by RefSeq
Specifications
CCR4 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of CCR4. | |
In PBS (0.09% sodium azide) | |
P51679 | |
CCR4 | |
A synthetic peptide (conjugated with KLH) corresponding to human CCR4. | |
50 μg | |
Primary | |
2nd extracellular domain of human. | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user. | |
CCR4 | |
CC-CKR-4/CD194/CKR4/CMKBR4/ChemR13/HGCN:14099/K5-5/MGC88293 | |
Rabbit | |
Immunoaffinity purification | |
RUO | |
1233 | |
Human, Monkey, Mouse, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only