Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR5 Rabbit anti-Human, Primates, Polyclonal Antibody, Abnova™

Catalog No. 89137512 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-512 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-512 Supplier Abnova Corporation Supplier No. PAB12576

Rabbit polyclonal antibody raised against synthetic peptide of CCR5.

Sequence: EWQKEGLVTLWLGGGGSC

Specifications

Antigen CCR5
Applications ELISA, Flow Cytometry, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation In antiserum
Host Species Rabbit
Immunogen A synthetic peptide corresponding to CCR5.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody is specific to CCR5. It demonstrates 2D7-like reactivity.
Target Species Human, Primate
Content And Storage Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.