Learn More
CCRL1 Rabbit anti-Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CCRL1.
Supplier: Abnova Corporation PAB26101
Description
The protein encoded by this gene is a member of the G protein-coupled receptor family, and is a receptor for C-C type chemokines. This receptor has been shown to bind dendritic cell- and T cell-activated chemokines including CCL19/ELC, CCL21/SLC, and CCL25/TECK. Alternatively spliced transcript variants encoding the same protein have been described. [provided by RefSeq
Specifications
CCRL1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of CCRL1. | |
In PBS (0.09% sodium azide) | |
Q9NPB9 | |
CCRL1 | |
A synthetic peptide corresponding to 16 amino acids at cytoplasmic domain of human CCRL1. | |
50 μg | |
Primary | |
BLAST analysis of the peptide immunogen showed no homology with other human proteins, except CCR6 (63%). | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (24 ug/mL) The optimal working dilution should be determined by the end user. | |
CCRL1 | |
CC-CKR-11/CCBP2/CCR10/CCR11/CCX-CKR/CKR-11/PPR1/VSHK1 | |
Rabbit | |
Immunoaffinity chromatography | |
RUO | |
51554 | |
Canine, Equine, Human, Mouse, Pig, Rabbit, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only