Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD14, Rabbit, Clone: SP192, Abnova™

Catalog No. 89110306 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-306 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-306 Supplier Abnova Corporation Supplier No. MAB12165

Rabbit monoclonal antibody raised against synthetic peptide of human CD14.

CD14 is a surface protein preferentially expressed on monocytes/macrophages. It binds lipopolysaccharide binding protein and recently has been shown to bind apoptotic cells. Alternative splicing results in multiple transcript variants encoding the same isoform. [provided by RefSeq

Specifications

Antigen CD14
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone SP192
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of human CD14.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) Western Blot (Tissue lysate) (1:100) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.6 (1% BSA, 0.09% sodium azide)
Gene CD14
Gene Symbols CD14
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus region of human CD14.
Purification Method Protein A/G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 929
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.