Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cd14 Rabbit anti-Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89117027 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-027 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-027 Supplier Abnova Corporation Supplier No. PAB14382
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant Cd14.

Specifications

Antigen Cd14
Applications ELISA, Flow Cytometry, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant Cd14.
Dilution Flow Cytometry (1:3000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene Cd14
Gene Symbols Cd14
Host Species Rabbit
Immunogen Recombinant protein corresponding to mouse Cd14.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12475
Test Specificity The antiserum represents a marker for mouse CD14. The CD14 glycoprotein, gp 55, is present on most monocytic and macrophages like cell types: The CD14 molecule has been reported to be a receptor for endotoxin.
Target Species Mouse
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.