Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD14 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89117026 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-026 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-026 Supplier Abnova Corporation Supplier No. PAB14381
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant CD14.

CD14 is a surface protein preferentially expressed on monocytes/macrophages. It binds lipopolysaccharide binding protein and recently has been shown to bind apoptotic cells. Alternative splicing results in multiple transcript variants encoding the same isoform. [provided by RefSeq

Specifications

Antigen CD14
Applications Flow Cytometry, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant CD14.
Dilution Flow Cytometry (1:100-1:1000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene CD14
Gene Symbols CD14
Host Species Rabbit
Immunogen Recombinant protein corresponding to human CD14.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 929
Test Specificity The antiserum represents a marker for human CD14. The CD14 glycoprotein, gp 55, is present on most monocytic and macrophages like cell types: monocytes, macrophages, Kupffer cells, pleural phagocytic cells and dendritic reticular cells. CD14 is also observed on granulocytes and activated or transformed B-cells. Furthermore CD14 is present in a soluble form in human serum, urine and other body fluids. The CD14 molecule has been reported to be a receptor for endotoxin.
Target Species Human, Mouse
Content And Storage Store at -80°C on dry atmosphere.
Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.