Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD19, Rabbit, Clone: SP110, Abnova™

Catalog No. 89108371 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-371 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-371 Supplier Abnova Corporation Supplier No. MAB10055

Rabbit monoclonal antibody raised against synthetic peptide of CD19.

Lymphocytes proliferate and differentiate in response to various concentrations of different antigens. The ability of the B cell to respond in a specific, yet sensitive manner to the various antigens is achieved with the use of low-affinity antigen receptors. This gene encodes a cell surface molecule which assembles with the antigen receptor of B lymphocytes in order to decrease the threshold for antigen receptor-dependent stimulation. [provided by RefSeq

Specifications

Antigen CD19
Applications Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone SP110
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of CD19.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.6 (1% BSA, 0.09% sodium azide)
Gene CD19
Gene Alias B4/MGC12802
Gene Symbols CD19
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human CD19.
Purification Method Protein A/G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 930
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.