Learn More
CD247 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CD247.
Supplier: Abnova Corporation PAB2493
Description
The protein encoded by this gene is T-cell receptor zeta, which together with T-cell receptor alpha/beta and gamma/delta heterodimers, and with CD3-gamma, -delta and -epsilon, forms the T-cell receptor-CD3 complex. The zeta chain plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways. Low expression of the antigen results in impaired immune response. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq
Specifications
CD247 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of CD247. | |
In PBS (0.09% sodium azide) | |
NP_932170;P20963 | |
CD247 | |
A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human CD247. | |
400 μL | |
Primary | |
Human | |
Liquid |
ELISA, Flow Cytometry, Western Blot | |
Unconjugated | |
Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
CD247 | |
CD3-ZETA/CD3H/CD3Q/CD3Z/T3Z/TCRZ | |
Rabbit | |
Protein A purification | |
RUO | |
919 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only