Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD5, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114944 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-944 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-944 Supplier Abnova Corporation Supplier No. PAB11231

Rabbit polyclonal antibody raised against synthetic peptide of CD5.

Specifications

Antigen CD5
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Concentration Lot specific
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CD5.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (Formalin/PFA-fixed paraffin-embedded sections) (1:200) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.4 (1% BSA, 0.09% sodium azide)
Gene CD5
Gene Alias LEU1/T1
Gene Symbols CD5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to intracellular region of human CD5.
Purification Method Immunoaffinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 921
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.